missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SLC7A1 (aa 604-629) Control Fragment Recombinant Protein

Numéro de catalogue. RP109653
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
RP109653 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. RP109653 Fournisseur Invitrogen™ Code fournisseur RP109653
Il en reste null

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (81%), Rat (81%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

SLC7A1 is a high-affinity, low capacity permease involved in the transport of the cationic amino acids (arginine, lysine and ornithine) in non-hepatic tissues. It may also function as an ecotropic retroviral leukemia receptor.

Spécifications

Accession Number P30825
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 6541
Name Human SLC7A1 (aa 604-629) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 4831426K01Rik; AI447493; amino acid transporter, cationic 1; ATRC1; Atrc-1; Cat1; CAT-1; cationic amino acid transporter 1; cationic amino acid transporter, y+ system; cationic amino acid transporter-1; cationic amion acid transporter; y+ system; cationinc amino acid transporter 1; ecotropic retroviral leukemia receptor; Ecotropic retroviral leukemia receptor homolog; ecotropic retroviral receptor; ecotropic retrovirus receptor; Ecotropic retrovirus receptor homolog; ERR; HCAT1; high affinity cationic amino acid transporter 1; mCAT-1; Rec-1; REC1L; Rev-1; Slc7a1; solute carrier family 7 (cationic amino acid transporter, y+ system), member 1; solute carrier family 7 member 1; Solute carrier family 7 member A1 (amino acid transporter cationic 1); solute carrier family 7, member 1; system Y+ basic amino acid transporter
Common Name SLC7A1
Gene Symbol SLC7A1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence GLWHSEEASLDADQARTPDGNLDQCK
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Afficher plus de résultats Afficher moins de résultats