missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SLC1A4 (aa 478-531) Control Fragment Recombinant Protein

Numéro de catalogue. RP102640
Modifier l'affichage
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
RP102640 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. RP102640 Fournisseur Invitrogen™ Code fournisseur RP102640

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (91%), Rat (91%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-111047 (PA5-111047. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Transporter for alanine, serine, cysteine, and threonine. Exhibits sodium dependence.

Spécifications

Accession Number P43007
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 6509
Name Human SLC1A4 (aa 478-531) Control Fragment
Quantity 100 μL
Gene Alias Alanine/serine/cysteine/threonine transporter 1; ASCT1; ASCT-1; AW045657; glutamate/neutral amino acid transporter; neutral amino acid transporter A; neutral amino acid transporter ASCT1; OTTHUMP00000159933; OTTHUMP00000235138; SATT; si:zc101n13.5; Slc1a4; slc1a4 protein; sodium-dependent neutral amino acid transporter ASCT1; solute carrier family 1 (glutamate/neutral amino acid transporter), member 4; solute carrier family 1 member 4; SPATCCM; zgc:92817
Common Name SLC1A4
Gene Symbol SLC1A4
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence NQKATKKGEQELAEVKVEAIPNCKSEEETSPLVTHQNPAGPVASAPELESKESV
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Afficher plus de résultats Afficher moins de résultats