missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human OPRD1 Control Fragment Recombinant Protein

Numéro de catalogue. RP108576
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
RP108576 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. RP108576 Fournisseur Invitrogen™ Code fournisseur RP108576
Il en reste null

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (71%), Rat (71%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

OPRD1 is an Opioid Receptor that is involved in the control of anxiety, depression, and pain. It is a promising target in the therapy of mood-altering disorders and drug addiction. Delta and kappa opioid receptors function synergistically by forming heterodimers. OPRD1 expression has been reported primarily in the brain. High OPRD1 expression has been documented in the cortex, caudate, hippocampus, nucleus accumbens, and putamen; low OPRD1 expression has been identified in the cerebellum, hypothalamus, substantia nigra, and thalamus. OPRD1 expression is reduced in the putamen and amygdala of Alzheimer's patients and in the putamen in Parkinson's disease.

Spécifications

Accession Number P41143
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 4985
Name Human OPRD1 Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias delta 1 opioid receptor; delta opioid receptor 1; delta-type opioid receptor; DOR; D-OR-1; DOR-1; K56; mDOR; MSL-2; Nbor; Opioid receptor; opioid receptor A; opioid receptor delta 1; opioid receptor, delta 1; OPRD; Oprd1; Ror-a
Common Name OPRD1
Gene Symbol OPRD1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LQPPLFANASDAYPSAFPSAGANASGPP
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Afficher plus de résultats Afficher moins de résultats