missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human NLRP6/NALP6 Alexa Fluor 350-conjugated Antibody, R&D Systems™
Description
NALP6 Monoclonal specifically detects NALP6 in Human samples. It is validated for Flow Cytometry (Intracellular Staining).
Spécifications
Spécifications
| Antigène | NALP6 |
| Applications | Flow Cytometry (Intracellular Staining) |
| Classification | Monoclonal |
| Clone | 920631 |
| Conjugué | Alexa Fluor 350 |
| Dilution | Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells |
| Formule | Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. |
| Numéro d’acquisition du gène | P59044 |
| Alias du gène | Angiotensin II/Vasopressin Receptor, AVR, CLR11.4, NACHT, Leucine Rich Repeat And PYD Containing 6, NACHT, LRR And PYD Containing Protein 6, NACHT, LRR And PYD Domains-Containing Protein 6, NALP6, NAVR, NAVR/AVR, NLR Family, Pyrin Domain Containing 6, NLRP6, Nucleotide-Binding Oligomerization Domain, Leucine Rich Repeat And Pyrin Domain Containing 6, PAN3, PYPAF5, PYRIN-Containing APAF1-Like Protein 5 |
| Espèces hôtes | Mouse |
| Afficher plus de résultats |
For Research Use Only (RUO)