missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MyD88 Alexa Fluor 405-conjugated Antibody, R&D Systems™
Description
MyD88 Monoclonal specifically detects MyD88 in Human samples. It is validated for Flow Cytometry (Intracellular Staining).
Spécifications
Spécifications
| Antigène | MyD88 |
| Applications | Flow Cytometry (Intracellular Staining) |
| Classification | Monoclonal |
| Clone | 316628 |
| Concentration | 0.2 mg/mL |
| Conjugué | Alexa Fluor 405 |
| Dilution | Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells |
| Formule | Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide |
| Numéro d’acquisition du gène | Q99836 |
| Alias du gène | MYD88D, myeloid differentiation primary response gene (88), myeloid differentiation primary response protein MyD88 |
| Afficher plus de résultats |
For Research Use Only (RUO)