missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat GSK-3 beta Alexa Fluor 488-conjugated Antibody, R&D Systems™
Description
GSK-3 beta Monoclonal specifically detects GSK-3 beta in Human∣Mouse∣Rat samples. It is validated for Flow Cytometry (Intracellular Staining).
Spécifications
Spécifications
| Antigène | GSK-3 beta |
| Applications | Flow Cytometry (Intracellular Staining) |
| Classification | Monoclonal |
| Clone | 272536 |
| Concentration | 0.2 mg/mL |
| Conjugué | Alexa Fluor 488 |
| Dilution | Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells |
| Formule | Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide |
| Numéro d’acquisition du gène | P49841 |
| Alias du gène | EC 2.7.11, EC 2.7.11.26, glycogen synthase kinase 3 beta, glycogen synthase kinase-3 beta, GSK3 beta, GSK-3 beta, GSK3B, GSK3beta isoform |
| Afficher plus de résultats |
For Research Use Only (RUO)