missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat GIT1 Alexa Fluor 700-conjugated Antibody, R&D Systems™
Description
GIT1 Monoclonal specifically detects GIT1 in Human∣Mouse∣Rat samples. It is validated for Flow Cytometry (Intracellular Staining).
Spécifications
Spécifications
| Antigène | GIT1 |
| Applications | Flow Cytometry (Intracellular Staining) |
| Classification | Monoclonal |
| Clone | 924640 |
| Conjugué | Alexa Fluor 700 |
| Dilution | Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells |
| Formule | Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. |
| Numéro d’acquisition du gène | Q9Y2X7 |
| Alias du gène | ARF GAP GIT1, ARF GTPase-activating protein GIT1, CAT1, CAT-1, Cool-associated and tyrosine-phosphorylated protein 1, G protein-coupled receptor kinase interacting ArfGAP 1, G protein-coupled receptor kinase interactor 1, G protein-coupled receptor kinase-interactor 1, GRK-interacting protein 1 |
| Espèces hôtes | Mouse |
| Afficher plus de résultats |
For Research Use Only (RUO)