missing translation for 'onlineSavingsMsg'
Learn More

Human/Mouse Pax3/Pax7 APC-conjugated Antibody, R&D Systems™

Numéro de catalogue. IC2457A
Modifier l'affichage
Click to view available options
Quantité:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
IC2457A 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. IC2457A Fournisseur Bio-Techne Code fournisseur IC2457A

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Mouse Monoclonal Antibody

Pax3/Pax7 Monoclonal specifically detects Pax3/Pax7 in Human∣Mouse samples. It is validated for Flow Cytometry (Intracellular Staining).

Spécifications

Antigène Pax3/Pax7
Applications Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone 274212
Conjugué APC
Dilution Intracellular Staining by Flow Cytometry 10 uL/10^6 cells
Formule Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Numéro d’acquisition du gène NP_000429
Alias du gène CDHS, HuP2, HUP2MGC120384, MGC120381, MGC120382, MGC120383, MGC134778, paired box 3, paired box gene 3 (Waardenburg syndrome 1), paired box homeotic gene 3, paired box protein Pax-3, paired domain gene 3, paired domain gene HuP2, Waardenburg syndrome 1, WS1, WS3
Symboles de gène PAX3
Espèces hôtes Mouse
Immunogène E. coli-derived recombinant human Pax3 (isoform Pax3a) Met1-Ser215 Accession # NP_000429
Méthode de purification Protein A or G purified from hybridoma culture supernatant
Quantité 100 Tests
Statut réglementaire RUO
Primaire ou secondaire Primary
ID du gène (Entrez) 5077
Spécifications de test Detects human and mouse Pax3 and Pax7 in direct ELISAs and Western blots. In direct ELISAs, approximately 20% cross-reactivity with recombinant human (rh) Pax1 is observed, approximately 10% with rhPax6, and less than 2% with rhPax2, rhPax4, rhPax5, and rhPax9.
Espèces cibles Human, Mouse
Contenu et stockage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Type de produit Antibody
Forme Purified
Isotype IgG2a
Gamme de produit R&D Systems
Afficher plus de résultats Afficher moins de résultats