missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human METTL12 (aa 164-234) Control Fragment Recombinant Protein
Description
Highest antigen sequence indentity to the following orthologs: Mouse (25%), Rat (25%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-58252 (PA5-58252. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Spécifications
Spécifications
| Numéro d'enregistrement | A8MUP2 |
| Concentration | ≥5.0 mg/mL |
| À utiliser avec (application) | Blocking Assay, Control |
| Formule | 1 M urea, PBS with no preservative; pH 7.4 |
| ID du gène (Entrez) | 751071 |
| Nom | Human METTL12 (aa 164-234) Control Fragment |
| Quantité | 100 μL |
| Statut réglementaire | RUO |
| Alias du gène | Citrate synthase-lysine N-methyltransferase CSKMT, mitochondrial; CSKMT; CS-KMT; methyltransferase like 12; methyltransferase-like protein 12, mitochondrial; METTL12; Putative methyltransferase LOC751071, mitochondrial; U99HG |
| Nom usuel | METTL12 |
| Afficher plus de résultats |