missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MDH1B (aa 1-72) Control Fragment Recombinant Protein

Numéro de catalogue. RP108624
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
RP108624 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. RP108624 Fournisseur Invitrogen™ Code fournisseur RP108624
Il en reste null

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (79%), Rat (79%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-84940 (PA5-84940. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

MDH1B is a protein coding gene.

Spécifications

Accession Number Q5I0G3
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 130752
Name Human MDH1B (aa 1-72) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias malate dehydrogenase 1 B; malate dehydrogenase 1 B, NAD (soluble); MDH1B; Putative malate dehydrogenase 1 B; RP11-95H11
Common Name MDH1B
Gene Symbol MDH1B
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence MAKFVIAGRADCPYYAKTELVADYLQKNLPDFRIHKITQRPEVWEDWLKDVCEKNKWSHKNSPIIWRELLDR
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Afficher plus de résultats Afficher moins de résultats