missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human lys-Plasminogen Native Protein

Catalog No. PIRP43081
Change view
Click to view available options
Quantité:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantité
PIRP43081 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIRP43081 Supplier Invitrogen™ Supplier No. RP43081

missing translation for 'validMainframeMsg'

Native Protein

RP-43081 contains native human lys-Plasminogen protein. RP-43081 may be used as a positive control in many immunological assays. The product is of human blood/plasma origin. Although the starting material was tested prior to initiation of the manufacturing process, and was found negative or nonreactive for anti-HIV-1/2, HIV-1 antigen(s), HBsAg, STS, anti-HCV, anti-HBcore and anti-HTLV I & II, extreme caution should be used when handling this material as there is a margin of error in all tests. This product is to be used for IN VITRO research purposes only, and is not intended for clinical or diagnostic use!.

Specifications

Numéro d'enregistrement P00747
Concentration 6.6 mg/mL
À utiliser avec (application) Neutralization
Formule 50% water with 50% glycerol and no preservative
ID du gène (Entrez) 5340
Poids moléculaire (g/mol) 83 kDa
Nom Human lys-Plasminogen
Méthode de purification SDS-PAGE
Quantité 1 mg
Sources Human
Conditions de stockage -20°C, Avoid Freeze/Thaw Cycles
Statut réglementaire RUO
Alias du gène Activation peptide; Angiostatin; plasmin; Plasmin heavy chain A; Plasmin heavy chain A, short form; Plasmin light chain B; Plasminogen; PLG
Nom usuel lys-Plasminogen
Symbole de gène PLG
Type de produit Protein
Conjugué Unconjugated
Espèces Human
Contenu et stockage -20°C, Avoid Freeze/Thaw Cycles
Forme Liquid
Sous-type de protéine Native
Show More Show Less