Learn More
Stemcell Technologies Inc Human LDL, Size: 5 mg
Numéro de catalogue. NC9055779
Encompass
This product is a human plasma-derived low-density lipoprotein (LDL)-based culture supplement that has been pre-screened for the culture, expansion, and colony assay of human hematopoietic and non-hematopoietic cells in serum-free culture media. It promotes the proliferation and survival of human hematopoietic and other progenitor cells in culture, resulting in increased cell output in expansion cultures and increased colony numbers and/or size in colony assays.
Numéro de catalogue. NC9055779 Fournisseur Stemcell Technologies Inc Code fournisseur 02698
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
This product is a human plasma-derived low-density lipoprotein (LDL)-based culture supplement that has been pre-screened for the culture, expansion, and colony assay of human hematopoietic and non-hematopoietic cells in serum-free culture media. It promotes the proliferation and survival of human hematopoietic and other progenitor cells in culture, resulting in increased cell output in expansion cultures and increased colony numbers and/or size in colony assays.