missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human KDELC2 (aa 442-500) Control Fragment Recombinant Protein
Description
Highest antigen sequence indentity to the following orthologs: Mouse (83%), Rat (83%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-84709 (PA5-84709. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
KDELC2 is a protein coding gene.
Spécifications
Spécifications
| Numéro d'enregistrement | Q7Z4H8 |
| Concentration | ≥5.0 mg/mL |
| À utiliser avec (application) | Blocking Assay, Control |
| Formule | 1 M urea, PBS with no preservative; pH 7.4 |
| ID du gène (Entrez) | 143888 |
| Nom | Human KDELC2 (aa 442-500) Control Fragment |
| Quantité | 100 μL |
| Statut réglementaire | RUO |
| Alias du gène | KDEL (Lys-Asp-Glu-Leu) containing 2; KDEL motif containing 2; KDEL motif-containing protein 2; KDELC2; POGLUT3; Protein O-glucosyltransferase 3; Protein O-xylosyltransferase POGLUT3; UNQ1904/PRO4350 |
| Nom usuel | KDELC2 |
| Afficher plus de résultats |