missing translation for 'onlineSavingsMsg'
Learn More

Human IL-6 Fluorescein-conjugated Antibody, R&D Systems™

Numéro de catalogue. IC206F
Modifier l'affichage
Click to view available options
Quantité:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
IC206F 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. IC206F Fournisseur Bio-Techne Code fournisseur IC206F

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Mouse Monoclonal Antibody has been used in 6 publications

IL-6 Monoclonal specifically detects IL-6 in Human∣Pig samples. It is validated for Flow Cytometry∣Flow Cytometry (Intracellular Staining).

Spécifications

Antigène IL-6
Applications Flow Cytometry, Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone 1936
Conjugué Fluorescein
Dilution Intracellular Staining by Flow Cytometry 10 uL/10^6 cells
Formule Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Numéro d’acquisition du gène P05231
Alias du gène B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL6, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6, MGI-2A
Symboles de gène IL6
Espèces hôtes Mouse
Immunogène E. coli-derived recombinant human IL-6 Val30-Met212 Accession # P05231
Méthode de purification Protein A or G purified from ascites
Quantité 100 Tests
Statut réglementaire RUO
Primaire ou secondaire Primary
ID du gène (Entrez) 3569
Spécifications de test Detects human IL-6 in direct ELISAs. Does not cross-react with recombinant IL-6 from mouse, rat, or pig.
Espèces cibles Human, Pig
Contenu et stockage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Type de produit Antibody
Forme Purified
Isotype IgG2b
Gamme de produit R&D Systems
Afficher plus de résultats Afficher moins de résultats