missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-2 Alexa Fluor™ 750-conjugated Antibody, R&D Systems™
Description
IL-2 Monoclonal specifically detects IL-2 in Human samples. It is validated for Flow Cytometry (Intracellular Staining).
Spécifications
Spécifications
| Antigène | IL-2 |
| Applications | Flow Cytometry (Intracellular Staining) |
| Classification | Monoclonal |
| Clone | 2171A |
| Conjugué | Alexa Fluor 750 |
| Dilution | Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells |
| Formule | Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. |
| Numéro d’acquisition du gène | P60568 |
| Alias du gène | aldesleukin, IL2, IL-2lymphokine, interleukin 2, interleukin-2, involved in regulation of T-cell clonal expansion, T cell growth factor, T-cell growth factor, TCGF |
| Espèces hôtes | Rabbit |
| Afficher plus de résultats |