Learn More
ACROBiosystems Inc resDetect™ Human Interleukin-2 (IL-2) ELISA Kit (Residue Testing)
Numéro de catalogue. 502719694
Encompass
Human Interleukin-2 (IL-2) ELISA Kit (Residue Testing) is based on ELISA sandwich method and designed to detect and quantitative determine of GMP human IL-2 in samples from CAR-T product preparation processing. It contains GMP human IL-2 and a pair of antibodies against the recombinant factor, which are provided by ACROBiosystems. Results are obtained by four parameter logistic curve that were parallel to the standard curves obtained. The verification results indicate that this kit can be used for the quantitative determination of GMP human IL-2 (ACROBiosystems, cat#GMP-L02H14) concentrations. The specificity has been verified.
Numéro de catalogue. 502719694 Fournisseur ACROBiosystems Inc Code fournisseur CRSA00396TESTS
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Human Interleukin-2 (IL-2) ELISA Kit (Residue Testing) is based on ELISA sandwich method and designed to detect and quantitative determine of GMP human IL-2 in samples from CAR-T product preparation processing. It contains GMP human IL-2 and a pair of antibodies against the recombinant factor, which are provided by ACROBiosystems. Results are obtained by four parameter logistic curve that were parallel to the standard curves obtained. The verification results indicate that this kit can be used for the quantitative determination of GMP human IL-2 (ACROBiosystems, cat#GMP-L02H14) concentrations. The specificity has been verified.