missing translation for 'onlineSavingsMsg'
Learn More

Human IL-17B Alexa Fluor 405-conjugated Antibody, R&D Systems™

Numéro de catalogue. IC1248V100
Modifier l'affichage
Click to view available options
Quantité:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
IC1248V100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. IC1248V100 Fournisseur Bio-Techne Code fournisseur IC1248V100UG

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Mouse Monoclonal Antibody

IL-17B Monoclonal specifically detects IL-17B in Human samples. It is validated for Flow Cytometry (Intracellular Staining).

Spécifications

Antigène IL-17B
Applications Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone 174113
Concentration 0.2 mg/mL
Conjugué Alexa Fluor 405
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formule Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Numéro d’acquisition du gène Q9UHF5
Alias du gène Cytokine Zcyto7, cytokine-like protein ZCYTO7, IL17B, IL-17Binterleukin-17 beta, IL20, IL-20interleukin-17B, interleukin 17B, interleukin 20, Interleukin-20, MGC138900, MGC138901, neuronal interleukin-17 related factor, Neuronal interleukin-17-related factor, NIRF, ZCYTO7interleukin-20
Symboles de gène IL17B
Espèces hôtes Mouse
Immunogène E. coli-derived recombinant human IL-17B Gln21-Phe180 Accession # Q9UHF5
Quantité 100 μg
Statut réglementaire RUO
Primaire ou secondaire Primary
ID du gène (Entrez) 27190
Spécifications de test Detects human IL-17B in Western blots and ELISAs. In Western blots, approximately 25% cross-reactivity with recombinant mouse IL-17B is observed and no cross-reactivity with recombinant human (rh) IL-17, rhIL-17C, rhIL-17E or rhIL-17F is observed. In sandwich immunoassays, less than 0.5% cross-reactivity with rmIL-17B, and no cross-reactivity with rhIL-17C, rhIL-17D, rhIL-17E, rhIL-17F, or rhIL-17 is observed.
Espèces cibles Human
Contenu et stockage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Type de produit Antibody
Isotype IgG2b
Gamme de produit R&D Systems
Afficher plus de résultats Afficher moins de résultats

For Research Use Only (RUO)