missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-10 Alexa Fluor 750-conjugated Antibody, R&D Systems™
Description
IL-10 Monoclonal specifically detects IL-10 in Human samples. It is validated for Flow Cytometry (Intracellular Staining).
Specifications
Specifications
| Antigène | IL-10 |
| Applications | Flow Cytometry (Intracellular Staining) |
| Classification | Monoclonal |
| Clone | 2050B |
| Conjugué | Alexa Fluor 750 |
| Dilution | Intracellular Staining by Flow Cytometry 0.25-1 μg/106 cells |
| Formule | Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. |
| Numéro d’acquisition du gène | P22301 |
| Alias du gène | CSIF, CSIFMGC126450, Cytokine synthesis inhibitory factor, GVHDS, IL10, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF |
| Espèces hôtes | Rabbit |
| Show More |