missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Inc Biotinylated Human IFN-alpha/beta R1 Protein (extracellular domain), C-terminal His and biotin tags - 25 g

Catalog No. 502370268
Encompass

Recombinant biotinylated human interferon alpha/beta receptor 1 (extracellular domain) expressed in HEK293 cells and supplied lyophilized. Designed for binding and analytical assays, the protein includes a C-terminal polyhistidine tag and an engineered biotinylation tag for enzymatic labeling; provided at high purity for SPR, MALS, and other characterization methods.

  • Expression system: HEK293 cells; amino acids Lys 28-Lys 436.
  • Biotinylated at an engineered tag site; includes a C-terminal His tag.
  • Calculated molecular weight ~50 kDa; migrates 66-90 kDa on reducing SDS-PAGE due to glycosylation.
  • Purity >90% as determined by SDS-PAGE and SEC-MALS.
  • Lyophilized from PBS, pH 7.4, with trehalose as protectant.
  • Available in a 25 g vial; suitable for SPR and other binding assays.

Catalog No. 502370268 Supplier ACROBiosystems Inc Supplier No. IF1H82E325UG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Recombinant biotinylated human interferon alpha/beta receptor 1 (extracellular domain) expressed in HEK293 cells and supplied lyophilized. Designed for binding and analytical assays, the protein includes a C-terminal polyhistidine tag and an engineered biotinylation tag for enzymatic labeling; provided at high purity for SPR, MALS, and other characterization methods.

  • Expression system: HEK293 cells; amino acids Lys 28-Lys 436.
  • Biotinylated at an engineered tag site; includes a C-terminal His tag.
  • Calculated molecular weight ~50 kDa; migrates 66-90 kDa on reducing SDS-PAGE due to glycosylation.
  • Purity >90% as determined by SDS-PAGE and SEC-MALS.
  • Lyophilized from PBS, pH 7.4, with trehalose as protectant.
  • Available in a 25 g vial; suitable for SPR and other binding assays.