missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Galanin Alexa Fluor 350-conjugated Antibody, R&D Systems™
Description
Galanin Monoclonal specifically detects Galanin in Human samples. It is validated for Flow Cytometry (Intracellular Staining).
Spécifications
Spécifications
| Antigène | Galanin |
| Applications | Flow Cytometry (Intracellular Staining) |
| Classification | Monoclonal |
| Clone | 581403 |
| Concentration | 0.2 mg/mL |
| Conjugué | Alexa Fluor 350 |
| Dilution | Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells |
| Formule | Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide |
| Numéro d’acquisition du gène | P22466 |
| Alias du gène | GAL, GAL1, galanin, galanin prepropeptide, GALN, GALNgalanin-related peptide, GLNNgalanin-message-associated peptide, GMAP, MGC40167 |
| Afficher plus de résultats |
For Research Use Only (RUO)