missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FGF-21 Alexa Fluor 700-conjugated Antibody, R&D Systems™
Description
FGF-21 Monoclonal specifically detects FGF-21 in Human samples. It is validated for Flow Cytometry (Intracellular Staining).
Spécifications
Spécifications
| Antigène | FGF-21 |
| Applications | Flow Cytometry (Intracellular Staining) |
| Classification | Monoclonal |
| Clone | 461804 |
| Concentration | 0.2 mg/mL |
| Conjugué | Alexa Fluor 700 |
| Dilution | Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells |
| Formule | Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide |
| Numéro d’acquisition du gène | Q9NSA1 |
| Alias du gène | FGF21, FGF-21, fibroblast growth factor 21 |
| Afficher plus de résultats |
For Research Use Only (RUO)