missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human FCHO1 (aa 285-364) Control Fragment Recombinant Protein
Description
Highest antigen sequence indentity to the following orthologs: Mouse (81%), Rat (81%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-83568 (PA5-83568. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
FCHO1 is involved in clathrin-coated vesicle formation.
Spécifications
Spécifications
| Numéro d'enregistrement | O14526 |
| Concentration | ≥5.0 mg/mL |
| À utiliser avec (application) | Blocking Assay, Control |
| Formule | 1 M urea, PBS with no preservative; pH 7.4 |
| ID du gène (Entrez) | 23149 |
| Nom | Human FCHO1 (aa 285-364) Control Fragment |
| Quantité | 100 μL |
| Statut réglementaire | RUO |
| Alias du gène | 3322402E17Rik; F-BAR domain only protein 1; FCH and mu domain containing endocytic adaptor 1; FCH domain only 1; FCH domain only protein 1; FCHO1; KIAA0290; N28152; unm_sa2424 |
| Nom usuel | FCHO1 |
| Afficher plus de résultats |