missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human EOMES Alexa Fluor 488-conjugated Antibody, R&D Systems™
Description
EOMES Monoclonal specifically detects EOMES in Human samples. It is validated for Flow Cytometry (Intracellular Staining).
Spécifications
Spécifications
| Antigène | EOMES |
| Applications | Flow Cytometry (Intracellular Staining) |
| Classification | Monoclonal |
| Clone | 644730 |
| Conjugué | Alexa Fluor 488 |
| Dilution | Intracellular Staining by Flow Cytometry 5 uL/10^6 cells |
| Formule | Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide |
| Numéro d’acquisition du gène | O95936 |
| Alias du gène | eomesodermin, eomesodermin (Xenopus laevis) homolog, t box, brain, 2, T-box brain protein 2, T-box brain2, TBR2, TBR-2, TBR2eomesodermin homolog, T-brain-2 |
| Symboles de gène | EOMES |
| Afficher plus de résultats |