missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Accession Number | 4932410A21Rik; CHD2; CHD2-42; CHD2-52; Down syndrome cell adhesion molecule; Down syndrome cell adhesion molecule homolog; DS cell adhesion molecule; Dscam |
|---|---|
| Assay Range | 1.4 to 300 ng/mL |
| Assay Sensitivity | 1.4 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
EH163RBX5
|
Invitrogen™
EH163RBX5 |
5 x 96 Tests |
Each for $4,420.00
|
|
|||||
EH163RB
|
Invitrogen™
EH163RB |
96 Tests |
Each for $890.00
|
|
|||||
Description
Human DSCAM quantitates human DSCAM in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human DSCAM.
This gene is a member of the immunoglobulin superfamily of cell adhesion molecules (Ig-CAMs), and is involved in human central and peripheral nervous system development. This gene is a candidate for Down syndrome and congenital heart disease (DSCHD). A gene encoding a similar Ig-CAM protein is located on chromosome 11. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene. [provided by RefSeq, Oct 2012].Specifications
| 4932410A21Rik; CHD2; CHD2-42; CHD2-52; Down syndrome cell adhesion molecule; Down syndrome cell adhesion molecule homolog; DS cell adhesion molecule; Dscam | |
| 1.4 ng/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| DSCAM | |
| <10 | |
| HRP | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| DSCAM | |
| 4 hrs 45 mins |
| 1.4 to 300 ng/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| 1826 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |