missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human DSCAM ELISA Kit

Catalog No. EH163RB
Encompass
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH163RB 96 Tests
EH163RBX5 5 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EH163RB Supplier Invitrogen™ Supplier No. EH163RB
Only null left

ELISA

Human DSCAM quantitates human DSCAM in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human DSCAM.

This gene is a member of the immunoglobulin superfamily of cell adhesion molecules (Ig-CAMs), and is involved in human central and peripheral nervous system development. This gene is a candidate for Down syndrome and congenital heart disease (DSCHD). A gene encoding a similar Ig-CAM protein is located on chromosome 11. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene. [provided by RefSeq, Oct 2012].

Specifications

Accession Number 4932410A21Rik; CHD2; CHD2-42; CHD2-52; Down syndrome cell adhesion molecule; Down syndrome cell adhesion molecule homolog; DS cell adhesion molecule; Dscam
Assay Range 1.4 to 300 ng/mL
Assay Sensitivity 1.4 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 1826
Gene Symbol DSCAM
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target DSCAM
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less