missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human CXCL4 (PF4) Recombinant Protein, PeproTech®

Numéro de catalogue. 3001620UG
Encompass
Modifier l'affichage
Click to view available options
Quantité:
100 μg
20 μg
250 μg
5 μg
50 μg
500 μg
1 mg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
3001620UG 20 μg
30016100UG 100 μg
300161MG 1 mg
30016250UG 250 μg
30016500UG 500 μg
3001650UG 50 μg
300165UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Numéro de catalogue. 3001620UG Fournisseur Gibco™ Code fournisseur LS3001620UG
Saison des économies
Afficher toutes les promotions

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Recombinant Protein

Recombinant Human PF-4 is a 7.8 kDa protein containing 70 amino acid residues, including the four highly conserved residues present in CXC chemokines. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Platelet factor-4 is a 70-amino acid protein that is released from the alpha-granules of activated platelets and binds with high affinity to heparin. Its major physiologic role appears to be neutralization of heparin-like molecules on the endothelial surface of blood vessels, thereby inhibiting local antithrombin III activity and promoting coagulation. As a strong chemoattractant for neutrophils and fibroblasts, PF4 probably has a role in inflammation and wound repair (Eisman et al., 1990).

Spécifications

Numéro d'enregistrement P02776
À utiliser avec (application) Control, Western Blot Control, Bioactivity
Formule Protein with no preservative
ID du gène (Entrez) 5196
Poids moléculaire (g/mol) 7.8 kDa
Nom Human CXCL4 (PF4)
Méthode de purification Purified
Quantité 20 μg
Sources E. coli
Conditions de stockage -20°C
Statut réglementaire RUO
Concentration en endotoxines <1 EU/μg
Alias du gène chemokine (C-X-C motif) ligand 4; C-X-C motif chemokine 4; Cxcl4; Endothelial cell growth inhibitor; Iroplact; oncostatin-A; PF4; PF-4; Pf4a; Platelet factor 4; Platelet factor 4, short form; RATPF4A; Scyb4
Nom usuel PF4
Symbole de gène Pf4
Activité biologique Determined by its ability to chemoattract human fibroblasts using a concentration range of 1.0-10.0 ng/ml
Type de produit Protein
Conjugué Unconjugated
Recombinant Recombinant
Séquence EAEEDGDLQC LCVKTTSQVR PRHITSLEVI KAGPHCPTAQ LIATLKNGRK ICLDLQAPLY KKIIKKLLES
Contenu et stockage -20°C
Activité Determined by its ability to chemoattract human fibroblasts using a concentration range of 1.0-10.0 ng/ml.
Système d’expression E. coli
Forme Lyophilized
Grade de qualité ou de pureté ≥ 98% by SDS-PAGE gel and HPLC analyses.
Afficher plus de résultats Afficher moins de résultats