missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human CLPH Synthetic Peptide
Description
This peptide corresponds to 15 amino acids near the carboxy terminus of human CLPH. PEP-0966 can be used as a blocking peptide with polyclonal antibody PA5-20852.
CLPH (Casein-Like phosphoprotein) is a recently discovered, evolutionarily conserved, testis-specific cytoplasmic protein expressed exclusively in postmeiotic germ cells in rat spermatogenesis. CLPH is a novel casein kinase 2-phosphorylated disordered and calcium-binding protein that displays strong variation in relative expression levels during spermatogenesis. It is frequently associated with the mitochondrial inner membrane during the last steps of spermatid differentiation and is involved in the extremely complex structural rearrangements occurring in haploid germ cells during spermiogenesis.
Spécifications
Spécifications
| Species | Human |
| Purification Method | purified |
| Accession Number | Q96KC9 |
| Common Name | CLPH |
| Concentration | 200 μg/mL |
| Format | Liquid |
| Formulation | PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 |
| Gene Alias | C4orf35; CABS1; calcium binding protein, spermatid associated 1; calcium-binding and spermatid-specific protein 1; calcium-binding protein, spermatid-specific 1; calcium-binding protein, sperm-specific 1; casein-like phosphoprotein; Clph; NYD-SP26; Protein RSD-6; Rsd6; RSD-6; Testis development protein NYD-SP26 |
| Gene Symbol | CABS1 |
| Lyophilized | No |
| Afficher plus de résultats |