Learn More
ACROBiosystems Inc FITC-Labeled Human CD99 / MIC2 Protein, Fc Tag
Numéro de catalogue. 502719795
Encompass
CD99 antigen is also known as MIC2, single-chain type-1 glycoprotein, HBA71, MIC2X, MIC2Y, MSK5X, 12E7, which belongs to the CD99 family. CD99 / MIC2 is expressed on all leukocytes but highest on thymocytes. Involved in T-cell adhesion processes and in spontaneous rosette formation with erythrocytes. CD99 / MIC2 plays a role in a late step of leukocyte extravasation helping leukocytes to overcome the endothelial basement membrane. CD99 acts at the same site as, but independently of PECAM1. There is also experimental evidence that it binds to cyclophilin A.
Numéro de catalogue. 502719795 Fournisseur ACROBiosystems Inc Code fournisseur CD9HF254200UG
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
CD99 antigen is also known as MIC2, single-chain type-1 glycoprotein, HBA71, MIC2X, MIC2Y, MSK5X, 12E7, which belongs to the CD99 family. CD99 / MIC2 is expressed on all leukocytes but highest on thymocytes. Involved in T-cell adhesion processes and in spontaneous rosette formation with erythrocytes. CD99 / MIC2 plays a role in a late step of leukocyte extravasation helping leukocytes to overcome the endothelial basement membrane. CD99 acts at the same site as, but independently of PECAM1. There is also experimental evidence that it binds to cyclophilin A.