Learn More
ACROBiosystems Inc Fluorescent Human CCR5 Full Length Protein (VLP)
Catalog No. 502719869
Encompass
Receptor for a number of inflammatory CC-chemokines including CCL3/MIP-1-alpha, CCL4/MIP-1-beta and RANTES and subsequently transduces a signal by increasing the intracellular calcium ion level. May play a role in the control of granulocytic lineage proliferation or differentiation.5 Publications (Microbial infection) Acts as a coreceptor (CD4 being the primary receptor) of human immunodeficiency virus-1/HIV-1.
Catalog No. 502719869 Supplier ACROBiosystems Inc Supplier No. CC5H52P5100UG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Receptor for a number of inflammatory CC-chemokines including CCL3/MIP-1-alpha, CCL4/MIP-1-beta and RANTES and subsequently transduces a signal by increasing the intracellular calcium ion level. May play a role in the control of granulocytic lineage proliferation or differentiation.5 Publications (Microbial infection) Acts as a coreceptor (CD4 being the primary receptor) of human immunodeficiency virus-1/HIV-1.