missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CCL3/MIP-1 alpha Alexa Fluor 700-conjugated Antibody, R&D Systems™
Description
CCL3/CCL4 Monoclonal specifically detects CCL3/CCL4 in Human samples. It is validated for Flow Cytometry (Intracellular Staining).
Spécifications
Spécifications
| Antigène | CCL3/CCL4 |
| Applications | Flow Cytometry (Intracellular Staining) |
| Classification | Monoclonal |
| Clone | 93342 |
| Concentration | 0.2 mg/mL |
| Conjugué | Alexa Fluor 700 |
| Dilution | Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells |
| Formule | Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide |
| Numéro d’acquisition du gène | P10147 |
| Alias du gène | C-C motif chemokine 3, MIP1-(a), AI323804, chemokine (C-C motif) ligand 3, G0S19-1, LD78a, LD78alpha, MIP1 alpha, MIP-1 alpha, Mip1a, MIP-1alpha, MIP1-alpha, Scya3 |
| Afficher plus de résultats |
For Research Use Only (RUO)