missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CCL22/MDC Alexa Fluor 750-conjugated Antibody, R&D Systems™
Description
CCL22/MDC Monoclonal specifically detects CCL22/MDC in Human samples. It is validated for Flow Cytometry (Intracellular Staining).
Spécifications
Spécifications
| Antigène | CCL22/MDC |
| Applications | Flow Cytometry (Intracellular Staining) |
| Classification | Monoclonal |
| Clone | 57203 |
| Concentration | 0.2 mg/mL |
| Conjugué | Alexa Fluor 750 |
| Dilution | Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells |
| Formule | Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide |
| Numéro d’acquisition du gène | O00626.1 |
| Alias du gène | A-152E5.1, ABCD-1, CC chemokine STCP-1, C-C motif chemokine 22, chemokine (C-C motif) ligand 22, DC/B-CK, Macrophage-derived chemokine, MDC, MDCStimulated T-cell chemotactic protein 1, MGC34554, SCYA22MDC(1-69), small inducible cytokine A22, small inducible cytokine subfamily A (Cys-Cys), member 22, Small-inducible cytokine A22, STCP-1, stimulated T cell chemotactic protein 1 |
| Afficher plus de résultats |
For Research Use Only (RUO)