missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human C2orf16 (aa 928-1013) Control Fragment Recombinant Protein
Description
Highest antigen sequence indentity to the following orthologs: Mouse (67%), Rat (67%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-62562. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Spécifications
Spécifications
| Numéro d'enregistrement | Q68DN1 |
| Concentration | ≥5.0 mg/mL |
| À utiliser avec (application) | Neutralization, Control |
| Formule | PBS, 1M urea with no preservative; pH 7.4 |
| ID du gène (Entrez) | 84226 |
| Nom | Human C2orf16 (aa 928-1013) Control Fragment |
| Plage de pH | 7.4 |
| Méthode de purification | Purified |
| Quantité | 100 μL |
| Conditions de stockage | -20°C, Avoid Freeze/Thaw Cycles |
| Afficher plus de résultats |