missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human C19orf47 (aa 277-354) Control Fragment Recombinant Protein
Description
Highest antigen sequence indentity to the following orthologs: Mouse (83%), Rat (83%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61210 (PA5-61210. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
C19orf47 (Chromosome 19 Open Reading Frame 47) is a Protein Coding gene.
Spécifications
Spécifications
| Numéro d'enregistrement | Q8N9M1 |
| Concentration | ≥5.0 mg/mL |
| À utiliser avec (application) | Blocking Assay, Control |
| Formule | 1 M urea, PBS with no preservative; pH 7.4 |
| ID du gène (Entrez) | 126526 |
| Nom | Human C19orf47 (aa 277-354) Control Fragment |
| Quantité | 100 μL |
| Statut réglementaire | RUO |
| Alias du gène | C19orf47; chromosome 19 open reading frame 47; uncharacterized protein C19orf47 |
| Nom usuel | C19orf47 |
| Afficher plus de résultats |