missing translation for 'onlineSavingsMsg'
Learn More

Genlantis™ Human Bronchial and Tracheal Epithelial Cells and Kits

Catalog No. PH50405A
Encompass
Change view
Click to view available options
Cell Type:
HBEC
HTEC
Quantity:
>500000 Cells
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Cell Type Quantity
PH50405A HTEC >500000 Cells
PH50205A HBEC >500000 Cells
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PH50405A Supplier Genlantis™ Supplier No. PH50405A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Human Bronchial/Tracheal Epithelial Cells (HBEpC/HTEpC), are derived from the surface epithelium of normal human bronchi. Cells are cryopreserved at first passage and can be cultured and propagated at least 16 population doublings. Airway epithelial cells are targets in asthma, viral infection, and fibrotic lung disease. These cells have been used to study the regulation of intracellular pH, the stimulant effect of IL-1b on airway epithelial cell growth, and the effect of human rhinovirus infection in terms of upregulation of ICAM-1 expression on epithelial cells.

Specifications

Cell Type HTEC
For Use With (Application) Used to study the regulation of intracellular pH, the stimulant effect of IL-1b on airway epithelial cell growth, the effect of human rhinovirus infection in terms of upregulation of ICAM-1 expression on epithelial cells
Media Recommendation PM511500, PM510500, PM11GS
Quantity >500000 Cells
Source Surface Epithelium of Normal Human Bronchi/Trachea
Sufficient For 16 Population Doublings