missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human alpha-Smooth Muscle Actin Alexa Fluor 488-conjugated Antibody, R&D Systems™
Description
alpha-Smooth Muscle Actin Monoclonal specifically detects alpha-Smooth Muscle Actin in Human∣Mouse∣Rat samples. It is validated for Flow Cytometry∣Flow Cytometry (Intracellular Staining)∣Immunohistochemistry.
Spécifications
Spécifications
| Antigène | alpha-Smooth Muscle Actin |
| Applications | Flow Cytometry, Flow Cytometry (Intracellular Staining), Immunohistochemistry |
| Classification | Monoclonal |
| Clone | 1A4 |
| Conjugué | Alexa Fluor 488 |
| Dilution | Intracellular Staining by Flow Cytometry 5 uL/10^6 cells |
| Formule | Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide |
| Alias du gène | AAT6, ACTA2, Actin alpha 2, actin, alpha 2, smooth muscle, aorta, actin, aortic smooth muscle, ACTSA, ACTVS, alpha 2 actin, alpha SMA, alpha smooth muscle actin, alpha-actin-2, alpha-SMA, alphaSmooth Muscle Actin, a-sma, Cell growth-inhibiting gene 46 protein, growth-inhibiting gene 46, MYMY5, sma, smooth muscle actin |
| Symboles de gène | ACTA2 |
| Espèces hôtes | Mouse |
| Afficher plus de résultats |