missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Adenylate Kinase 9 (aa 581-668) Control Fragment Recombinant Protein

Numéro de catalogue. RP94911
Modifier l'affichage
Click to view available options
Quantité:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
RP94911 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. RP94911 Fournisseur Invitrogen™ Code fournisseur RP94911

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (59%), Rat (59%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-56564 (PA5-56564. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Involved in maintaining the homeostasis of cellular nucleotides by catalyzing the interconversion of nucleoside phosphates. Has both nucleoside monophosphate and diphosphate kinase activities. Catalyzes the phosphorylation of AMP, dAMP, CMP and dCMP with ATP as phosphate donor and of CMP with GTP as phosphate donor. Also catalyzes the production of ATP, CTP, GTP, UTP, dATP, dCTP, dGTP and TTP from the corresponding diphosphate substrates with either ATP or GTP as phosphate donor. Shows substrate preference of CDP - UDP - ADP - GDP - TDP.

Spécifications

Numéro d'enregistrement Q5TCS8
Concentration ≥5.0 mg/mL
À utiliser avec (application) Blocking Assay, Control
Formule 1 M urea, PBS with no preservative; pH 7.4
ID du gène (Entrez) 221264
Nom Human Adenylate Kinase 9 (aa 581-668) Control Fragment
Quantité 100 μL
Statut réglementaire RUO
Alias du gène Adenylate kinase 9; adenylate kinase domain containing 1; adenylate kinase domain containing 2; Adenylate kinase domain-containing protein 1; Adenylate kinase domain-containing protein 2; AK 9; AK9; AKD1; AKD2; C6orf199; C6orf224; dJ70A9.1; Gm234; Gm7127; RGD1560386; RP1-70A9.1; similar to novel protein
Nom usuel Adenylate Kinase 9
Symbole de gène AK9
Conjugué Unconjugated
Espèces Human
Recombinant Recombinant
Étiquette de la protéine His-ABP-tag
Séquence VVTMIEETIKMSQDINFEQPYEKHAEILQEVLGEVMEENKDRFPGAPKYGGWIVDNCPIVKELWMALIKKGIIPDLVIYLSDTENNGK
Contenu et stockage -20°C, Avoid Freeze/Thaw Cycles
Système d’expression E. coli
Forme Liquid
Grade de qualité ou de pureté >80% by SDS-PAGE and Coomassie blue staining
Afficher plus de résultats Afficher moins de résultats