missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ABCA1 Alexa Fluor 750-conjugated Antibody, R&D Systems™
Description
ABCA1 Monoclonal specifically detects ABCA1 in Human samples. It is validated for Flow Cytometry (Intracellular Staining).
Spécifications
Spécifications
| Antigène | ABCA1 |
| Applications | Flow Cytometry (Intracellular Staining) |
| Classification | Monoclonal |
| Clone | 768510 |
| Concentration | 0.2 mg/mL |
| Conjugué | Alexa Fluor 750 |
| Dilution | Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells |
| Formule | Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide |
| Numéro d’acquisition du gène | O95477 |
| Alias du gène | ABC1, ABC-1, ABC1ATP-binding cassette transporter A1, ATP-binding cassette 1, ATP-binding cassette transporter 1, ATP-binding cassette, sub-family A (ABC1), member 1, CERP, Cholesterol efflux regulatory protein, EC 2.7.7.8, EC 2.8.1.8, EC 3.6.3, EC 3.6.3.41, FLJ14958, HDLDT1, membrane-bound, MGC164864, MGC165011, TGD, TGDATP-binding cassette sub-family A member 1 |
| Afficher plus de résultats |
For Research Use Only (RUO)