missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HSPA12A Recombinant Rabbit Monoclonal Antibody (23GB6030)
GREENER_CHOICE

Numéro de catalogue. MA552974
Modifier l'affichage
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
MA552974 100 μL
MA552975 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. MA552974 Fournisseur Invitrogen™ Code fournisseur MA552974
Il en reste null

Rabbit Recombinant Monoclonal Antibody

HSPA12A Recombinant Monoclonal Antibody for ICC/IF, Western Blot

HSPA12A is a protein coding gene.

Spécifications

Antigen HSPA12A
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB6030
Concentration 1.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene HSPA12A
Gene Accession No. O43301, Q8K0U4
Gene Alias 1700063D12Rik; AI118035; AI840429; AW048913; AW556406; D5Mgi40; heat shock 70 kDa protein 12A; heat shock 70kD protein 12A; heat shock 70kDa protein 12A; Heat shock protein; heat shock protein 12A; heat shock protein family A (Hsp70) member 12A; HSP; HSPA12A; KIAA0417; mKIAA0417
Gene Symbols HSPA12A
Host Species Rabbit
Immunogen A synthesized peptide derived from human HSPA12A (104-143AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 259217, 73442
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Afficher plus de résultats Afficher moins de résultats