missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ HP35 Disposable Microtome Blades

Catalog No. 28618168
Change view
Click to view available options
Coating:
Coated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Coating
28618168 Coated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 28618168 Supplier Epredia™ Supplier No. 3150734

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Epredia™ HP35 microtome blades are coated for reliable and thin sectioning.

  • High-profile
  • Increased durability

Specifications

Product Type High Profile Blade
Profile High
For Use With (Application) Designed for high-quality thin sectioning
Disposable Single-use
Coating Coated
Cutting Angle 34°
Thickness (English) 0.01 in.
Thickness (Metric) 0.31 mm