Learn More
SP Scientific Hotpack Glassware Washer Accessories
Accessories designed for use with Hotpack™ Undercounter, Freestanding and Vertical Stacking Glassware washers safeguard flasks, beakers, test tubes, bottles, Petri dishes, and more.
$1,077.89 - $6,428.07
Specifications
For Use With (Application) Washing Lab Glassware
View More Specs
Products 21
Catalog Number Mfr. No. Product Type Dimensions (L x W x H) Holds Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Dimensions (L x W x H) Holds Price Quantity & Availability  
15365224
View Documents Promotion Details
SP Scientific
437127
Basket and Cover 7.5 in. x 7.5 in. x 4 in. General use: small glassware
Each for $2,248.71
Only null left
15365227
View Documents Promotion Details
SP Scientific
437158
Petri Dish Basket Insert 24 in. x 12 in. x 4 in.
Each for $2,929.46
Only null left
15365230
View Documents Promotion Details
SP Scientific
437189
Test Tube Basket Insert with Lid 12 in. x 12 in. x 6 in. Up to 150, 75mm long test tubes
Each for $3,783.18
Only null left
15365231
View Documents Promotion Details
SP Scientific
437196
Test Tube Basket Insert with Lid 12 in. x 12 in. x 4 in. Up to 150, 105mm long test tubes
Each for $6,428.07
Only null left
15365232
View Documents Promotion Details
SP Scientific
437202
Test Tube Basket Insert with Lid 12 in. x 12 in. x 4 in. Up to 150, 165mm long test tubes
Each for $4,949.39
Only null left
15365233
View Documents Promotion Details
SP Scientific
437219
Test Tube Basket Insert with Lid 12 in. x 12 in. x 4 in. Up to 150, 200mm long test tubes
Each for $5,831.00
Only null left
15365234
View Documents Promotion Details
SP Scientific
437226
Open Basket Insert with Handles 12 in. x 12 in. x 4 in. General use: small glassware
Each for $4,112.39
Only null left
15365236
View Documents Promotion Details
SP Scientific
437240
Bottle Basket Insert 24 in. x 24 in. x 6 in. Up to 16, 1L bottles
Each for $2,566.75
Only null left
15365237
View Documents Promotion Details
SP Scientific
437257
Bottle Basket Insert 24 in. x 24 in. x 6 in. Up to 9, 2L bottles
Each for $2,243.14
Only null left
15365238
View Documents Promotion Details
SP Scientific
437264
Bottle Basket Insert 24 in. x 24 in. x 6' Up to 69, 50mL bottles
Each for $2,198.50
Only null left
15365239
View Documents Promotion Details
SP Scientific
437271
Bottle Basket Insert 24 in. x 24 in. x4 in. Up to 40, 100mL bottles
Each for $3,124.75
Only null left
15365240
View Documents Promotion Details
SP Scientific
437288
Bottle Basket Insert 24 in. x 24 in. x 4 in. Up to 24, 250mL bottles
Each for $3,191.71
Only null left
15365241
View Documents Promotion Details
SP Scientific
437295
Bottle Basket Insert 24 in. x 24 in. x 4 in. Up to 23, 500mL bottles
Each for $1,891.61
Only null left
15365244
View Documents Promotion Details
SP Scientific
437332
Basket Cover
Each for $1,077.89
Only null left
15365225
View Documents Promotion Details
SP Scientific
437134
Erlenmeyer Flask Insert, 300 to 1500 mL 24 in. x 12 in. x 6 in. Up to 5 Erlenmeyer flasks
Each for $1,305.71
Only null left
15365226
View Documents Promotion Details
SP Scientific
437141
BOD Bottle Insert 24 in. x 12 in. x 4 in. Up to 18 BOD bottles
Each for $1,495.43
Only null left
15365229
View Documents Promotion Details
SP Scientific
437172
Arrow Clip Rack Insert 24 in. x 12 in. x 12 in. Miscellaneous glassware on 14 small and 14 large arrow clips
Each for $3,587.89
Only null left
15365235
View Documents Promotion Details
SP Scientific
437233
Watch Glass Insert 24 in. x 24 in. x 4 in. Up to 40 pieces less than 125mm in dia.
Each for $1,573.54
Only null left
15365242
View Documents Promotion Details
SP Scientific
437301
Bottom Mesh Insert 24 in. x 24 in.
Each for $2,304.50
Only null left
15365243
View Documents Promotion Details
SP Scientific
437318
Bottom Mesh Insert 12 in. x 24 in.
Each for $2,204.07
Only null left
15365218
View Documents Promotion Details
SP Scientific
446508
Booster Pump
Each for $3,694.57
Only null left
Description
Description
  • Accessories maximize space, improve efficiency, and protect specialty glassware; use in standard open racks
  • Bottom mesh inserts prevent small items from falling through
  • Made of stainless steel for long life and durability
  • Booster Pump for use with low pressure DI water supply below 18psi
Specifications
Specifications
Washing Lab Glassware