missing translation for 'onlineSavingsMsg'
Learn More

Troemner™ Hooked Stainless-Steel Weight Sets and Individual Weights

Numéro de catalogue. p-3750640
Change view
Click to view available options
Product Type:
Metric Hook Weight
Weight (Metric):
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Product Type Weight (Metric)
S40304A Metric Hook Weight 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. S40304A Fournisseur Troemner™ Code fournisseur 30390790
Il en reste null

Weights can be hooked together at top and bottom.

The design of the hook weight allows the bottom hook to be recessed in the bottom so the hook weights can be set on a flat surface.

  • Adjusted to tolerances specified by ANSI™/ASTM™ Class 7
  • Weight sets are supplied in an open plastic storage case

Spécifications

Product Type Metric Hook Weight
Classification ANSI/ASTM Class 7
Material Stainless Steel
Weight (Metric) 50 g
Density 7.85g/cm3 at 20°C