missing translation for 'onlineSavingsMsg'
Learn More

Honokiol 95.0+%, TCI America™

Supplier:  TCI America H1309200MG

 View more versions of this product

Catalog No. H1309200MG


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Honokiol
85°C
MFCD00016674
honokiol, 5,3'-diallyl-2,4'-dihydroxybiphenyl, 3,5'-diallyl-4,2'-dihydroxybiphenyl, unii-11513cco0n, chembl16901, 3',5-diallyl-1,1'-biphenyl-2,4'-diol, 4-allyl-2-3-allyl-4-hydroxy-phenyl phenol, 3',5-diallyl-2,4'-biphenyldiol, 3',5-diallylbiphenyl-2,4'-diol, 5,3′-diallyl-2,4′-dihydroxydiphenyl
C=CCC1=CC(=C(C=C1)O)C2=CC(=C(C=C2)O)CC=C
266.34
CHEBI:5759
≥95.0% (GC)
35354-74-6
C18H18O2
200 mg
FVYXIJYOAGAUQK-UHFFFAOYSA-N
2-(4-hydroxy-3-prop-2-enylphenyl)-4-prop-2-enylphenol
72303
266.34
Crystalline Powder

Chemical Identifiers

35354-74-6
266.34
FVYXIJYOAGAUQK-UHFFFAOYSA-N
72303
2-(4-hydroxy-3-prop-2-enylphenyl)-4-prop-2-enylphenol
C18H18O2
MFCD00016674
honokiol, 5,3'-diallyl-2,4'-dihydroxybiphenyl, 3,5'-diallyl-4,2'-dihydroxybiphenyl, unii-11513cco0n, chembl16901, 3',5-diallyl-1,1'-biphenyl-2,4'-diol, 4-allyl-2-3-allyl-4-hydroxy-phenyl phenol, 3',5-diallyl-2,4'-biphenyldiol, 3',5-diallylbiphenyl-2,4'-diol, 5,3′-diallyl-2,4′-dihydroxydiphenyl
CHEBI:5759
C=CCC1=CC(=C(C=C1)O)C2=CC(=C(C=C2)O)CC=C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions