accessibility menu, dialog, popup

UserName

CAS RN 35354-74-6

IUPAC Name: 3',5-bis(prop-2-en-1-yl)-[1,1'-biphenyl]-2,4'-diol
Synonyms: honokiol
Molecular Weight (g/mol): 266.34
Molecular Formula: C18H18O2
InChi Key: FVYXIJYOAGAUQK-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1CC=C)C1=CC(CC=C)=CC=C1O
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)