missing translation for 'onlineSavingsMsg'
Learn More

Honokiol 95.0+%, TCI America™

Catalog No. p-7130971
Change view
Click to view available options
Quantity:
200 mg
1 g
Catalog No. Quantity
H1309200MG 200 mg
H13091G 1 g
2 options

Catalog No. H1309200MG

Supplier: TCI America H1309200MG

Only null left

Chemical Identifiers

CAS 35354-74-6
Molecular Formula C18H18O2
Molecular Weight (g/mol) 266.34
MDL Number MFCD00016674
InChI Key FVYXIJYOAGAUQK-UHFFFAOYSA-N
Synonym honokiol, 5,3'-diallyl-2,4'-dihydroxybiphenyl, 3,5'-diallyl-4,2'-dihydroxybiphenyl, unii-11513cco0n, chembl16901, 3',5-diallyl-1,1'-biphenyl-2,4'-diol, 4-allyl-2-3-allyl-4-hydroxy-phenyl phenol, 3',5-diallyl-2,4'-biphenyldiol, 3',5-diallylbiphenyl-2,4'-diol, 5,3′-diallyl-2,4′-dihydroxydiphenyl
PubChem CID 72303
ChEBI CHEBI:5759
IUPAC Name 2-(4-hydroxy-3-prop-2-enylphenyl)-4-prop-2-enylphenol
SMILES C=CCC1=CC(=C(C=C1)O)C2=CC(=C(C=C2)O)CC=C

Specifications

Melting Point 85°C
Quantity 200 mg
Formula Weight 266.34
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Honokiol
TSCA No
Recommended Storage Refrigerator