missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ Pure Flow 1000 Portable Emergency Eyewash Station

Catalog No. 18999265A
Change view
Click to view available options
Capacity (English):
892.6 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
18999265A 892.6 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18999265A Supplier Honeywell™ Supplier No. 320010000000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Vinyl eyewash sign, 8 x 10 in. (20.3 x 25.4cm)

Requires

Two solution cartridges (Cat. No. 18-999-265B, sold separately).

Self-contained, gravity-fed emergency eyewash

Delivers 7 gal. (26.4L) saline solution—purified, contaminant-free and physiologically correct—from a sealed, integrated delivery module.

  • Highly visible green color provides easy identification in an emergency situation Sure-grip handle provides instant, simple activation Activation straps display expiration date
  • Integral drain valve allows easy draining of reservoir after use
  • Integral fluid reservoir captures waste fluid, eliminating potential safety hazards and simplifying fluid disposal Cover keeps dirt and trash from collecting in the basin and nozzle
  • Unit can be serviced in five minutes or less
  • With provision for locking device
  • Saline eyewash cartridges are sold separately; see 320010500000H5

Compliance

Constant flowrate meets ANSI™ Z358.1-2004.

Specifications

Capacity (English) 892.6 oz.
Capacity (Metric) 26397.33 mL
Certifications/Compliance ANSI Z358.1-2004
Depth (English) 17.25 in.
Depth (Metric) 43.81 cm
Height (English) 30 in.
Height (Metric) 76.2 cm
Includes Vinyl Eyewash sign, mount-on-wall bracket, two cartridges with solution, delivery tube, nozzle assemblies
Mounting Wall
Product Type Eyewash Station