missing translation for 'onlineSavingsMsg'
Learn More

Homophthalic Acid 99.0+%, TCI America™

Catalog No. T026225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T026225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T026225G Supplier TCI America Supplier No. T026225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 89-51-0
Melting Point 181°C
Color White-Yellow
Molecular Formula C9H8O4
Quantity 25 g
MDL Number MFCD00004326
Synonym homophthalic acid, 2-carboxymethyl benzoic acid, benzeneacetic acid, 2-carboxy, 2-carboxyphenylacetic acid, 2-carboxybenzeneacetic acid, alpha-carboxy-o-toluic acid, o-carboxymethyl benzoic acid, 2-carboxyphenyl acetic acid, o-toluic acid, alpha-carboxy, toluene-alpha,2-dicarboxylic acid
InChI Key ZHQLTKAVLJKSKR-UHFFFAOYSA-N
SMILES C1=CC=C(C(=C1)CC(=O)O)C(=O)O
IUPAC Name 2-(carboxymethyl)benzoic acid
Molecular Weight (g/mol) 180.159
PubChem CID 66643
Formula Weight 180.16
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Homophthalic Acid
Show More Show Less
EINECSNumber (3)-1700
RTECSNumber CY1575590
TSCA Yes