missing translation for 'onlineSavingsMsg'
Learn More

Homogentisic Acid 97.0+%, TCI America™

Catalog No. D10505G
Change view
Click to view available options
Quantity:
200 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D10505G 5 g
D1050200MG 200 mg
D10501G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D10505G Supplier TCI America Supplier No. D10505G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 451-13-8
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.15
MDL Number MFCD00004324
InChI Key IGMNYECMUMZDDF-UHFFFAOYSA-N
Synonym homogentisic acid, 2,5-dihydroxyphenylacetic acid, alcapton, homogentisate acid, homogentisinic acid, 2,5-dihydroxyphenyl acetic acid, homogentisate, benzeneacetic acid, 2,5-dihydroxy, 2-2,5-dihydroxyphenyl acetic acid, 2,5-dihydroxy-alpha-toluic acid
PubChem CID 780
ChEBI CHEBI:44747
IUPAC Name 2-(2,5-dihydroxyphenyl)acetic acid
SMILES OC(=O)CC1=CC(O)=CC=C1O

Specifications

Melting Point 147°C
Color White-Yellow
Quantity 5 g
Formula Weight 168.15
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Homogentisic Acid