missing translation for 'onlineSavingsMsg'
Learn More

Cobaltous Chloride TS, (U.S.P. Test Solution), Spectrum™ Chemical

Catalog No. 18601788
Change view
Click to view available options
Quantity:
100 mL
1 L
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601788 1 L Amber Glass Bottle
18601787 100 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18601788 Supplier Spectrum Chemical Mfg Cor Supplier No. C2891LT
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

C-289, 7791-13-1

Spectrum™ Chemical Cobaltous Chloride TS, (U.S.P. Test Solution) is a solution, sometimes designated with 'TS or Test Solution', that is prepared according to the specifications of the USP Pharmacopeial Convention, for the purpose of conducting USP and National Formulary tests and assays.

Chemical Identifiers

CAS 7791-13-1 , 7732-18-5
Molecular Formula Cl2CoH12O6
Molecular Weight (g/mol) 237.92
MDL Number MFCD00149652
InChI Key GFHNAMRJFCEERV-UHFFFAOYSA-L
IUPAC Name λ2-cobalt(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Co++]

Specifications

CAS Max % 0.1%, 99.9%
Grade Chemical Solution
Packaging Amber Glass Bottle
Quantity 1 L
Chemical Name or Material Cobaltous Chloride TS