missing translation for 'onlineSavingsMsg'
Learn More
Learn More
hnRNP-U Polyclonal Antibody, Bethyl Laboratories
Description
The recommended shelf life for this product is 1 year from date of receipt.
The protein encoded by this gene is a receptor for activated C-kinase (RACK) protein. The encoded protein has been shown to bind in vitro to activated protein kinase C beta I. In addition, this protein is a cutaneous T-cell lymphoma-associated antigen. Finally, the protein contains a bromodomain and two zinc fingers, and is thought to be a transcriptional regulator. Multiple transcript variants encoding several different isoforms have been found for this gene.
Spécifications
Spécifications
| Antigène | hnRNP-U |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugué | Unconjugated |
| Formule | TBS with 0.1% BSA and 0.09% sodium azide |
| Gène | HNRNPU |
| Numéro d’acquisition du gène | Q00839, Q8VEK3 |
| Alias du gène | C1orf199, chromosome 1 open reading frame 199, FLJ30202, FLJ37978, heterogeneous nuclear ribonucleoprotein U (scaffold attachment factor A), hnRNP U, HNRNPU antisense RNA 1, HNRNPU antisense RNA 1 (non-protein coding), HNRNPU-AS1, HNRPU, NCRNA00201, non-protein coding RNA 201, p120, p120 nuclear protein, pp120, SAFA, SAF-A, Scaffold attachment factor A, U21.1 |
| Symboles de gène | HNRNPU |
| Espèces hôtes | Rabbit |
| Afficher plus de résultats |