missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ HM 430, HM 450 and 440 E Sliding Microtome Accessories

Numéro de catalogue. p-4529875
Change view
Click to view available options
Product Type:
Blade Holder, hi-profile blade
Blade Holder, lo-profile blade
Brain Rsrch Freezing Platform
Height Adjustment Plate, 10 mm
Height Adjustment Plate, 20 mm
Height Adjustment Plate, 5 mm
Std Specimen Clamp w/ Tray
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Product Type
361013368 Height Adjustment Plate, 10 mm
361013369 Height Adjustment Plate, 20 mm
361013370 Height Adjustment Plate, 5 mm
705200 Blade Holder, lo-profile blade
361013409 Blade Holder, hi-profile blade
361013443 Std Specimen Clamp w/ Tray
981000 Brain Rsrch Freezing Platform
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Numéro de catalogue. 361013368 Fournisseur Epredia™ Code fournisseur 440400
Il en reste null

Maintain Epredia™ HM 430 and HM 450 sliding microtomes with Epredia™ HM 430, HM 450 and 440 E sliding microtomes accessories.

Standard buffer tray

  • Holds stages in place using a magnet
  • Fill tray with all water-based buffers and solutions
  • Cooling element for buffer solution

  • Use pre-cooled in the standard buffer tray for cooling tissue
  • Use frozen together with buffer solution to prevent the frozen buffer solution from inflating
  • Large-field magnifier

  • Magnification 2.5X
  • Move, swivel, and displace for optimal viewing position
  • Includes adapter and illumination
  • StemiScope™ with optics and mount

  • Eyepieces: 10X
  • Supplementary lens: 0.63X
  • Zoom magnifying range: 4.1 to 31.5X
  • Microscope adapter for angled view of specimen
  • Horizontal view and vertical drive and swivel mechanism
  • Swan-neck light guide

  • 3.5 mm fiber bundle
  • Swan-neck light guide: 50 mm long
  • Flexible light guide: 750 mm long
  • High light intensity
  • Include adapter for the KL200 (760317)
  • Spécifications

    Product Type Height Adjustment Plate, 10 mm
    For Use With (Equipment) Epredia HM 430 and HM 450 Sliding Microtomes